Order Entry
Netherlands
ContactUsLinkComponent
2-Acetylfuran 99%
Toxic
2-Acetylfuran 99%
Certificates
undefined
Toxic
2-Acetylfuran 99%

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5060 to 1.5080 (20°C, 589 nm)
  • GC:
    ≤0.2 % 2,5-Diacetyl furan

Specification Test Results

Related Information

Frequently Bought Together