Order Entry
Netherlands
Orders LinkContactUsLinkComponent
1,1,1-Tris(hydroxymethyl)ethane ≥97%

1,1,1-Tris(hydroxymethyl)ethane ≥97%

1,1,1-Tris(hydroxymethyl)ethane ≥97%
Certificates
1,1,1-Tris(hydroxymethyl)ethane ≥97%
1,1,1-Tris(hydroxymethyl)ethane ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 77-85-0 Molecular Formula: C5H12O3 Formula Weight: 120.15 Flash Point: 160°(320°F) Melting Point: ca 190° Boiling Point: 137°/15mm Beilstein Reference: 1304452 MDL No.: MFCD00004687

Synonyms:  2-(Hydroxymethyl)-2-methyl-1,3-propanediol, Ethylidynetrimethanol
  • CAS Number:77-85-0
  • Formula:C₅H₁₂O₃
  • MDL Number:MFCD00004687
  • Molecular Weight:120.15 g/mol
  • Boiling Pt:137 °C (15 mmHg)
  • Melting Pt:∼ 190 °C
  • Flash Pt:160 °C (320 °F)
  • Storage Temperature:Ambient
  • EINECS:201-063-9
Product Family Options

Product Information