4'-Methylacetophenone
Supplier: CHEMPUR
Certificates
About this item
CAS Number:122-00-9
:
Formula:C₉H₁₀O
MDL Number:MFCD00008751
:134.18 g/mol
Boiling Pt:226 °C (1013 hPa)
Melting Pt:28 °C
Density:1.013 g/cm³ (20 °C)
Flash Pt:92 °C
EINECS:204-514-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together