Organic reference standard, 4-Chloro-m-cresol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11440300ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:59-50-7
- Description:4-Chloro-m-cresol 100 µg/ml in methanol
- Formula:C₇H₇ClO
- Description:4-Chloro-m-cresol