5-Chloro-2-methoxybenzonitrile
Catalog # MAYBTL00283.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
About this item
:
:167.59 g/mol
UN:3439
CAS Number:55877-79-7
Formula:C₈H₆ClNO
MDL Number:MFCD00052867
Boiling Pt:276
Melting Pt:99
ADR:6.1 III