Organic reference standard, o-Toluidine 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA17594800CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-53-4
- Description:o-Toluidine 100 µg/ml in cyclohexane
- Formula:C₇H₉N
- Description:o-Toluidine