Order Entry
Netherlands
ContactUsLinkComponent
Trimesic acid trimethyl ester 99%
Trimesic acid trimethyl ester 99%
Certificates
undefined
Trimesic acid trimethyl ester 99%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    143 to 147 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together