Order Entry
Netherlands
ContactUsLinkComponent
1,4-Dithioerythritol, Millipore®
Irritant
1,4-Dithioerythritol, Millipore®
Catalog # 233152-5
CAS Number:  
Irritant
1,4-Dithioerythritol, Millipore®
Catalog # 233152-5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :154.25 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:82…83 °C
EINECS:229-998-8
 :GHS07
Storage Temperature:Ambient
CAS Number:6892-68-8
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00063750