Acetonitrile ≥99.8% (by GC), BAKER ANALYZED®, Isocratic Grade for HPLC, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
Boiling Pt:81.6
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Molecular Weight:41.05 g/mol
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
UN:1648
ADR:3 II
Merck Index:13,00071
EINECS:200-835-2
REACH:01-2119471307-38
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (by GC):min. 99.8 %
- Residue after Evaporation:max. 0.0005 %
- Water (H2O):max. 0.1 %
- at 200 nm:max. 0.5
- at 210 nm:max. 0.3
- at 220 nm:max. 0.2
- at 254 nm:max. 0.05
- at 280 nm:max. 0.01
- UV Cut-off, nm:max. 190
Specification Test Results
Related Information
Certificates
Search by Lot NumberVWR J.T.Baker® High Purity solvents and reagents
Product Literature
Frequently Bought Together