Order Entry
Netherlands
ContactUsLinkComponent
Bromonitromethane 90% tech.
Irritant
Oxidizing
Bromonitromethane 90% tech.
Catalog # ACRO307050100
CAS Number:  
undefined
Irritant
Oxidizing
Bromonitromethane 90% tech.
Catalog # ACRO307050100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4850 to 1.4950 (20°C, 589 nm)
  • GC:
    ≥88 %

Specification Test Results

About this item

 :
 :139.94 g/mol
Boiling Pt:146…148 °C (750 mmHg)
Melting Pt:–28 °C
Density:2.07 g/cm³
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:209-258-0
UN:3139
ADR:5.1,II
 :GHS07
 :GHS03
CAS Number:563-70-2
Formula:BrCH₂NO₂
MDL Number:MFCD00007401