1-Bromononane for synthesis, Sigma-Aldrich®
Catalog # 8.21710.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Molar Mass:207.15 g/mol
- Boiling Point:201 °C (1013 hPa)
- Flash Point:90 °C
- Refractive Index:1.454 (20 °C, 589 nm)
- Density:1.09 g/cm3 (20 °C)
- Synonyms:Nonyl bromide
- Hill Formula:C9H19Br
- Chemical Formula:CH3(CH2)8Br
Specification Test Results
About this item
:
:207.15 g/mol
Boiling Pt:201 °C (1013 hPa)
Melting Pt:–29 °C
Density:1.086 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:211-755-2
CAS Number:693-58-3
Formula:BrH₂C(CH₂)₇CH₃
MDL Number:MFCD00000278