
1-Bromononane 97%
:693-58-3
:
:BrH₂C(CH₂)₇CH₃
:MFCD00000278
:207.15 g/mol
:201 °C (760 mmHg)
:-29 °C
:1.08 g/cm³
:90 °C
:211-755-2
:GHS07
:Ambient
- Appearance:Clear colorless to golden Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4550 (20°C, 589 nm)
- GC:≥96.0 %
- Absorption Ratio P-15/P+15:0.96 to 1.22
- BSC Certification:Passes
Frequently Bought Together