
Strontium chloride hexahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 28321.268
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- pH (20°C; 5 %):5.0 - 7.0
- Insolubility in water:Max. 50 ppm
- NO3 (Nitrate):Max. 20 ppm
- SO4 (Sulphate):Max. 25 ppm
- Ba (Barium):Max. 50 ppm
- Ca (Calcium):Max. 0.05 %
- Cu (Copper):Max. 2 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 100 ppm
- Na (Sodium):Max. 150 ppm
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 2 ppm
Specification Test Results
About this item
:
:266.62 g/mol
Merck Index:13,08920
Boiling Pt:100 °C (1013 hPa)
Melting Pt:61 °C
Density:1.93 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-971-6
:GHS07
CAS Number:10025-70-4
Formula:SrCl₂·6H₂O
MDL Number:MFCD00149865