Order Entry
Netherlands
ContactUsLinkComponent
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Specification Test Results
  • Titer (mol/l):
    0.0997 - 0.1003
  • Heavy Metals (as Pb):
    max. 1
  • Sulfate and Sulfite (as SO4):
    max. 50