Order Entry
Netherlands
ContactUsLinkComponent
Hydrochloric acid 0.5 mol/l (0.5 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
Hydrochloric acid 0.5 mol/l (0.5 N), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
 :  Avantor
  • Titer (mol/l):
    0.4975 - 0.5025
  • Ammonium (NH4):
    max. 3
  • Iron (Fe):
    max. 0.1
  • Heavy Metals (as Pb):
    max. 0.5
  • Sulfate (SO4):
    max. 1
Frequently Bought Together