Order Entry
Netherlands
ContactUsLinkComponent
 
Sulphuric acid ≥90-91%, Supelco®
 :  Merck
 
Sulphuric acid ≥90-91%, Supelco®
 :  Merck
 

 

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Boiling Point:
    300 °C
  • Density:
    1.81 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) strongly acid

 

Frequently Bought Together