Hydrogen peroxide 30%
VWRS84538.410
:
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
:
:∼107 °C (1013 hPa)
:< 0 °C
:1.11 g/cm³ (20 °C)
:5.1,II
:34.01 g/mol
:231-765-0
:2014
:GHS05
:GHS07
:Ambient
:7722-84-1
:H₂O₂
:MFCD00011333