Nitric acid 68%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8 (5.1),II
:GHS03
:GHS05
:GHS06
Specification Test Results
- Assay:68.0 - 70.0 %
- Density (20/4):1.404 - 1.413
- Ignition residue (SO4):Max. 10 ppm
- Cl (Chloride):Max. 0.5 ppm
- PO4 (Phosphate):Max. 2 ppm
- SO4 (Sulphate):Max. 2 ppm
- As (Arsenic):Max. 0.01 ppm
- Fe (Iron):Max. 1 ppm
- Mn (Manganese):Max. 0.4 ppm
- Pb (Lead):Max. 0.05 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
20422_NITRIC ACID 68 %_BSE_REV0
BSE/TSE Certificates
spec_VWRC20422.242 spec_VWRC20422.242 spec_VWRC20422.297 spec_VWRC20422.297 spec_VWRC20422.320
Product specification
More
Frequently Bought Together