Hydrogen peroxide 27% (w/w) in aqueous solution stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Merck Index:14,4798
Boiling Pt:107 °C
Melting Pt:–26 °C
Density:1.100
Storage Temperature:Ambient
EINECS:231-765-0
ADR:5.1,II
:34.01 g/mol
UN:2014
:GHS03
:GHS05
:GHS07
Related Information
Frequently Bought Together