Order Entry
Netherlands
ContactUsLinkComponent
2-Methyl-2-pentenal 97%
Flammable
Corrosive
Toxic
2-Methyl-2-pentenal 97%
Certificates
undefined
Flammable
Corrosive
Toxic
2-Methyl-2-pentenal 97%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4470 to 1.4550 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together