1,4-Dichloro-2-butene
Catalog # MOLEM49664205
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:125 g/mol
Boiling Pt:156 °C (1013 hPa)
Melting Pt:–47.8 °C
Density:1,1882 g/cm³ (20 °C)
EINECS:212-121-8
:GHS06
:GHS05
:GHS08
:GHS09
CAS Number:764-41-0
Formula:C₄H₆Cl₂
MDL Number:MFCD00000988
Melting Pt:–47,8 °C