
Calcium ≥99.98% (metals basis), crystalline dendritic pieces
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-70-2
:
Formula:Ca
MDL Number:MFCD00085314
:40.08 g/mol
Merck Index:13,01644
Boiling Pt:1484 °C (1013 hPa)
Melting Pt:850 °C
Density:1.54 g/cm³ (20 °C)
UN:1401
ADR:4.3,II
:ampouled under argon
EINECS:213-179-5
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together