Order Entry
Netherlands
ContactUsLinkComponent
 
Nitric acid 65%
  VWRS84537.360
 :  
Nitric acid 65%
  VWRS84537.360
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

 

 

 :
 :63.01 g/mol
 :13,06608
 :120.5 °C (1013 hPa)
 :–42 °C
 :1.4 g/cm³ (20 °C)
 :Ambient
 :231-714-2
 :2031
 :8,II
 :GHS03
 :GHS05
 :GHS06
 :7697-37-2
 :HNO₃
 :MFCD00011349