Order Entry
Netherlands
ContactUsLinkComponent
Xylène Cyanole
Irritant
Xylène Cyanole
Catalog # WAKO242-00462
Supplier:  Wako
CAS Number:  
Irritant
Xylène Cyanole
Catalog # WAKO242-00462
Supplier:  Wako
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :538.62 g/mol
Melting Pt:295 °C
EINECS:220-167-5
 :GHS07
Storage Temperature:Ambient
CAS Number:2650-17-1
Formula:C₂₅H₂₇N₂NaO₆S₂
MDL Number:MFCD00040657