Order Entry
Netherlands
ContactUsLinkComponent
Penicillinase (beta-Lactamase)
Penicillinase (beta-Lactamase)
Catalog # CHMCAB3738
Supplier:  Chemicon
CAS Number:  
Penicillinase (beta-Lactamase)
Catalog # CHMCAB3738
Supplier:  Chemicon
CAS Number:  

Specifications

  • Pk:
    100 µl
  • CAS No.:
    9001-74-5
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Penicillinase (beta-Lactamase)

Specifications

About this item