Order Entry
Netherlands
ContactUsLinkComponent
Pig Pancreatin, Protease, Lipase, Amylase (from Pancreas)
Pig Pancreatin, Protease, Lipase, Amylase (from Pancreas)
  1.07130.1000
 :  
Pig Pancreatin, Protease, Lipase, Amylase (from Pancreas)
  1.07130.1000
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • CAS No.:
    8049-47-6
  • Source:
    Pancreas
  • Species:
    Pig
  • Seq:
    01
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Pancreatin, Protease, Lipase, Amylase
  • Enzyme activity:
    Pancreatin 350 FIP-U/g Protease, 6000 FIP-U/g Lipase, 7500 FIP-U/g Amylase