Lithium acetate hydrate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:546-89-4
:
Formula:H₃CCOOLi·xH₂O
MDL Number:MFCD00013057
:65.98 g/mol (anhy)
Merck Index:14,5521
Melting Pt:291 °C
Density:1.26 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-914-3
Frequently Bought Together