Indigo carmine solution Reag. Ph. Eur. 1045601
Catalog # 87852.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:466.36 g/mol
EINECS:212-728-8
:GHS05
CAS Number:860-22-0
Formula:C₁₆H₈N₂Na₂O₈S₂
MDL Number:MFCD00005723