
Indigo carmine, Supelco®
Catalog # 1.04724.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:10 g/l (25 °C)
- Molar Mass:466.36 g/mol
- Bulk Density:700 - 900 kg/m3
- pH:5.3 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:466.36 g/mol
Merck Index:13,04969
Melting Pt:> 300 °C
:M-Clarity™ quality level = MQ200
EINECS:212-728-8
:GHS07
Storage Temperature:Ambient
CAS Number:860-22-0
Formula:C₁₆H₈N₂Na₂O₈S₂
MDL Number:MFCD00005723