Order Entry
Netherlands
ContactUsLinkComponent
Schiff reagent for analysis of aldehydes
Health
Schiff reagent for analysis of aldehydes
Catalog # ROTHN057.1
Supplier:  Roth Carl
CAS Number:  
Health
Schiff reagent for analysis of aldehydes
Catalog # ROTHN057.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
Density:1.1 g/cm³ (20 °C)
 :GHS08
MDL Number:MFCD00081985