Order Entry
Netherlands
ContactUsLinkComponent
Wide horizontal gel electrophoresis systems, Owl™ D2
Wide horizontal gel electrophoresis systems, Owl™ D2
Catalog # 730-0221
undefined
Wide horizontal gel electrophoresis systems, Owl™ D2
Catalog # 730-0221
Specifications
  • Description:
    Owl D2-BP complete system with buffer exchange ports
  • Buffer volume:
    600 ml
  • Gel size W×L:
    144×102 mm
  • Unit W×L×H:
    200×190×10 mm
  • Pk:
    1
Specifications
About this item
The Owl™ D2 wide gel system is ideal for screening PCR samples or for routine DNA fragment analysis.
  • Possible to run 10 to 80 samples in one gel
  • Wide gel size allows loading of 15 or 30 samples per row with a multi-channel pipette
  • EasyCast UV-transmissible gel tray fits snugly into the external gel caster to provide a leak-proof seal without tape
  • Multi-channel pipette compatible combs speed up sample transfer and reduce the chance of sample order error
  • Unique electrode placement allows the gel to run quickly without compromising the resolution
* Optimal for multi-channel pipetting.
Delivery: Each system includes buffer chamber, SuperSafe lid with attached power leads, EasyCast UV-transmissible gel tray with gasketed end gates, external multiple gel caster, and four combs: 1,5 mm thick, two 30-well, two 40-well.
Combs
Product Information