Wipes for general applications, Sontara® AC™
Catalog # 115-0371
Supplier: CONTEC
Specifications
- Packed:100 per pop-up box, 8 boxes per case
- L×W:419×229 mm
- Pk:800
Specifications
About this item
Launderable non-woven wipes made from DuPont™ Sontara AC fabric. Designed for aircraft manufacturing, these wipes offer benefits to numerous manufacturing applications particularly cleaning and polishing bright metallic surfaces. Interleaved polyester/cellulose blend with creped texture and a basis weight of 74,6 g/m². No metal particles.
- Highly absorbent, low-linting for snag resistance
- Able to withstand many solvents without falling apart
- Manufactured with no contaminating binders or glue
- Wipes come in pop-up boxes for easy dispensing