Gas inlet/air bleed tubes
Duran® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Straight inlet tube with cone, that can be cut to required length or drawn as a capillary for an air/gas bleed.
Useful length: 200 mm
Frequently Bought Together