Order Entry
Netherlands
ContactUsLinkComponent
Scalpel handles, B3/B3L
Scalpel handles, B3/B3L
Catalog # 233-5461
Supplier:  SWANN MORTON
undefined
Scalpel handles, B3/B3L
Catalog # 233-5461
Supplier:  SWANN MORTON
Specifications
  • For:
    Blades No. 6/9/10/10A/11/11P/E/11/12/12D/13/14/15/15A/15C/D/15/15T/16/40 and SG3
  • Type:
    B3
  • Length:
    127 mm
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Stainless steel, non sterile.
  • Smooth tapered octagonal body offers precise fingertip control
  • Suitable for hand surgery such as a Fasciectomy (removal of scar like tissue ), peripheral nerve repair and surgery of the tendons and joints
  • B3L is longer than the standard B3 providing greater reach and depth of surgical field