Order Entry
Netherlands
ContactUsLinkComponent
Transfer pipettes, micro
Transfer pipettes, micro
Catalog # 612-1761
Supplier:  Alpha Laboratories
undefined
Transfer pipettes, micro
Catalog # 612-1761
Supplier:  Alpha Laboratories

Specifications

  • Packed:
    Bag
  • Sterile:
    -
  • Capacity:
    3,0 ml
  • Length:
    88 mm
  • Drop vol.:
    40 µl
  • Pk:
    500

Specifications

About this item

Non toxic, food grade LDPE.

  • For small diameter tubes and cuvettes
  • Can be heat sealed for transport