Order Entry
Netherlands
Orders LinkContactUsLinkComponent
VWR®, Pasteur Pipettes
VWR®, Pasteur Pipettes
VWR®, Pasteur Pipettes
VWR®, Pasteur Pipettes
Soda-lime glass, disposable.
  • Non sterile
  • Automated manufacturing process ensures total production uniformity
  • Tapered upper opening for cotton ball plugging
  • Top body outer diameter: 7,1 mm, wall thickness: 0,53 mm, jet outer Ø: 1,5 mm
Product Family Options
  • Description
    Capacity
    Length
  • Non plugged
    2 ml
    150 mm
    1 * 250 ST
    1 * 250 ST - 17,60 €
    17,60 €
    1 * 250 ST
       17,60 €
  • Non plugged
    2 ml
    230 mm
    1 * 250 ST
    1 * 250 ST - 19,30 €
    19,30 €
    1 * 250 ST
       19,30 €
  • Non plugged, sealed tip, breakable
    2 ml
    230 mm
    1 * 250 ST
    1 * 250 ST - 19,80 €
    19,80 €
    1 * 250 ST
       19,80 €
  • Non plugged, sealed tip, breakable
    2 ml
    270 mm
    1 * 250 ST
    1 * 250 ST - 30,80 €
    30,80 €
    1 * 250 ST
       30,80 €
  • Pre-plugged
    2 ml
    150 mm
    1 * 250 ST
    1 * 250 ST - 28,60 €
    28,60 €
    1 * 250 ST
       28,60 €
  • Pre-plugged
    2 ml
    230 mm
    1 * 250 ST
    1 * 250 ST - 29,90 €
    29,90 €
    1 * 250 ST
       29,90 €
  • Pre-plugged, sealed tip, breakable
    2 ml
    230 mm
    1 * 250 ST
    1 * 250 ST - 26,80 €
    26,80 €
    1 * 250 ST
       26,80 €
  • Pre-plugged, sealed tip, breakable
    2 ml
    270 mm
    1 * 250 ST
    1 * 250 ST - 36,50 €
    36,50 €
    1 * 250 ST
       36,50 €