Order Entry
Netherlands
ContactUsLinkComponent
HTS 24-multiwell insert system, Falcon®
HTS 24-multiwell insert system, Falcon®
Supplier:  Corning
   Sustainable Options Available
Certificates
undefined
HTS 24-multiwell insert system, Falcon®
Supplier:  Corning
   Sustainable Options Available
Certificates

Specifications

  • Catalog No:
  • 734-0379
  • 734-0383
  • Effective diameter of membrane:
  • 6,5 mm
  • 6,5 mm
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Effective growth area of membrane:
  • 0,3 cm²
  • 0,3 cm²
  • Effective growth area in 24-well plate:
  • 2,0 cm²
  • 2,0 cm²
  • Distance from membrane to bottom of well:
  • 2,0 mm
  • 2,0 mm
  • Insert height:
  • 18 mm
  • 18 mm
  • Suggested media volume in insert:
  • 300 - 500 µl
  • 300 - 500 µl
  • Suggested media volume in well:
  • 1000 - 4000 µl
  • 1000 - 4000 µl
  • Environmental certification:
  • ISO 14001 - Bedford

Specifications

Related Information

Frequently Bought Together