Membrane filter funnels, magnetic
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Polyphenylsulfone, autoclavable.
- Leakproof magnetic seal allows one-handed operation
- Compatible with anti-foaming agents and many other solvents
For membrane filtration techniques for water testing.
Effective filtration area: 150 and 300 ml: 9,6 cm²; 500 ml: 13,1 cm²
Overall Ø×H: 150 ml: 76×178 mm; 300 ml: 76×229 mm; 500 ml: 89×196 mm
Related Information
Pall Laboratory Microbiology QC Products
Product Literature
PALL4241_TDS_EN PALL4242_TDS_EN
Datasheet
Frequently Bought Together