Order Entry
Netherlands
ContactUsLinkComponent
Tangential flow filtration systems and capsules, Minimate™ EVO
undefined
Tangential flow filtration systems and capsules, Minimate™ EVO
  • Catalog No:
  • 516-1085
  • Effective filtration area:
  • 50 cm2
  • Dimensions:
  • 307×482×208 mm
  • Pressure:
  • 4.1 bar (410 kPa, 60 psi)
Tangential flow filtration capsules
Frequently Bought Together