Cable Reels, 4-Way
Supplier: BACHMANN ELEKTRO
Certificates
About this item
Cable reel with four Schuko outlets 250 V~, without lid. Stable zinc-plated stand with black PVC cable (HO5VV-F G3 1,50 mm²).
- Design of side wheels helps dissipate heat and humidity
- Integrated one pole thermal cut-off switch
- Right angle plug with double earth
- VDE compliant
Colour: Black.
Max. power: Fully unrolled 3500 W; cable on reel 1000 W.
Frequently Bought Together