Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
Crucibles
Crucibles, quartz
Crucibles, quartz
:
NEUBERT VOLUME GLASSWAERE
Crucibles, quartz
:
NEUBERT VOLUME GLASSWAERE
Transparent quartz glass.
Withstand constant maximum temperature of 1100 °C, short term use at 1300 °C
Lids are available separately.
Lids