Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, LiChrolut®
Solid phase extraction cartridges, LiChrolut®
Solid phase extraction cartridges, LiChrolut®
Supplier:  Merck
Certificates
Solid phase extraction cartridges, LiChrolut®
Solid phase extraction cartridges, LiChrolut®
8 Options Available Below
Product Details & Documents
LiChrolut® solid phase extraction (SPE) cartridges and accessories are manufactured to the highest standards. All SPE cartridges listed below are polypropylene unless otherwise stated. They are compatible with all automated SPE instruments and robots. Three important criteria are carefully controlled.
  • Batch-to-batch reproducibility - each sorbent is tested for capacity towards polar and non polar compounds
  • Elimination of fines -  sorbents have a narrow particle size distribution with virtually no fines
  • Contamination - high purity sorbents and tubes are used and are pre-washed prior to packing ensuring contamination-free products
EN - For non polar extraction on a polymeric phase
SCX- For cation exchange extraction
TSC- For mixed mode analysis of neutral and cationic analytes
* For ultra-trace analysis
Product Family Options
Product Information
  • Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • 1.19127.0001
    LiChrolut® Florisil®
    LiChrolut® Florisil®
    Florisil®
    150 - 250 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 153,00 €
    153,00 €
    1 * 30 ST
       153,00 €
  • 1.19855.0001
    LiChrolut® RP18
    LiChrolut® RP18
    RP18
    40 - 63 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 413,00 €
    413,00 €
    1 * 100 ST
       413,00 €
  • 1.02014.0001
    LiChrolut® RP18
    LiChrolut® RP18
    RP18
    40 - 63 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 308,00 €
    308,00 €
    1 * 50 ST
       308,00 €
  • 1.02023.0001
    LiChrolut® RP18
    LiChrolut® RP18
    RP18
    40 - 63 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 461,00 €
    461,00 €
    1 * 50 ST
       461,00 €
  • LiChrolut® RP18
    RP18
    40 - 63 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 550,00 €
    550,00 €
    1 * 30 ST
       550,00 €
  • 1.02122.0001
    LiChrolut® RP18
    LiChrolut® RP18
    RP18
    40 - 63 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 541,95 €
    541,95 €
    1 * 30 ST
       541,95 €
  • 1.19847.0001
    LiChrolut® RP18E
    LiChrolut® RP18E
    RP18E
    40 - 63 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 301,00 €
    301,00 €
    1 * 50 ST
       301,00 €
  • 1.19849.0001
    LiChrolut® RP18E
    LiChrolut® RP18E
    RP18E
    40 - 63 µm
    3 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 553,00 €
    553,00 €
    1 * 50 ST
       553,00 €