Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®
   Sustainable Options Available
Dropping bottles, LDPE, Nalgene®
   Sustainable Options Available
Certificates
Dropping bottles, LDPE, Nalgene®
3 Options Available Below
Product Details & Documents
LDPE, transparent with colour-coded (red, green, blue, white and yellow) PP screw caps and LDPE dosing tip.
  • Good chemical resistance, heat resistant up to 80 °C
  • Drop control tip snaps securely into place, drops are dispensed individually
  • Flexible for easy handling, leakproof
  • Delivers 40 µl drops (based on water; viscosity affects drop size)
Reagent identification is very easy with the varied colours of the screw caps. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.
Specifications for Product Family
Specifications
  • Catalog No:
  • 215-1063
  • 215-1061
  • 215-1062
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 4 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 52,30 €
    52,30 €
    1 * 25 ST
       52,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1063
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 8 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 54,70 €
    54,70 €
    1 * 25 ST
       54,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1061
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 15 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 56,90 €
    56,90 €
    1 * 25 ST
       56,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1062
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact