Bottles, narrow neck, round, LDPE or PP, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
LDPE, translucent or PPCO, translucent, with PP screw cap.
- LDPE: Good chemical resistance/PPCO: Very good chemical resistance
- LDPE: Heat resistant up to 80 °C/PPCO: Heat resistant up to 121 °C, autoclavable
- Leakproof
Large volume bottles for shipping or storing chemicals, standard solutions, distilled water etc. The 4000 and 8000 ml sizes incorporate a loop for attaching a label.
Specifications
- Catalog No:
- 215-7744
- 215-7748
- 215-7742
- 215-7758
- 215-7752
- 215-7754
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement