Order Entry
Netherlands
ContactUsLinkComponent
VWR®, Blotting Pads, Grade 707
undefined
VWR®, Blotting Pads, Grade 707
  • Catalog No:
  • 732-0608
  • 732-0606
  • 732-0601
  • 732-0604
  • 732-0602
  • 732-0594
  • 732-0599
  • 732-0595
  • 732-0600
  • 732-0592
  • 732-0597
  • 732-0598
  • Grade:
  • 707
  • 707
  • 707
  • 707
  • 707
  • 707
  • 707
  • 707
  • 707
  • 707
  • 707
  • 707
  • Weight:
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • 703 g/m²
  • Thickness:
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • 2,60 mm
  • Herzberg flow rate:
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • 120 s per 100 ml at 10 cm head of water
  • Klemm wicking rate test:
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
  • 96 s per 7,5 cm rise
Frequently Bought Together