Quinaldine red, Supelco®
Catalog # 1.02282.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:240 °C (decomposition)
- Molar Mass:430.33 g/mol
- Bulk Density:290 kg/m3
Specification Test Results
About this item
:
:430.33 g/mol
Merck Index:12,08230
Melting Pt:240 °C
:M-Clarity™ quality level = MQ200
EINECS:204-221-5
CAS Number:117-92-0
Formula:C₂₁H₂₃IN₂
MDL Number:MFCD00011968