Alizarin complexone ∼85% for the determination of fluorine, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:3952-78-1
:
Formula:C₁₉H₁₅NO₈
MDL Number:MFCD00001202
:385.33 g/mol
Melting Pt:185 °C
Storage Temperature:Ambient
EINECS:223-544-2
Specification Test Results
- Appearance:complying
- Assay:80.0 - 120.0 %
- Appearance of the solution:complying
- Loss on drying (110°C):Max. 10 %
- Absorption maximum:422 - 432 nm
- Absorbtion (1%/1cm):Min. 75
Specification Test Results
Frequently Bought Together