Methyl cellulose, 4000 mPa.S (2% H₂O)
:9004-67-5
:
:MFCD00081763
:Ambient
- Appearance:White to yellowish-white to greyish-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Viscosity:3000 to 5600 cP (2 % in water at 20°C)
Frequently Bought Together