Methyl cellulose, 1500 mPa.S (2% H₂O), fine white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9004-67-5
:
MDL Number:MFCD00081763
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Viscosity (2% aq soln, 25°C):1125 - 2100 cps
- Loss on Drying :<5.0%
Specification Test Results
Frequently Bought Together