Order Entry
Netherlands
ContactUsLinkComponent
Human/Mouse/Rat Recombinant GDF-11 (from E. coli)
Human/Mouse/Rat Recombinant GDF-11 (from E. coli)
Certificates
Human/Mouse/Rat Recombinant GDF-11 (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Rat
    Mouse
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: Alkaline phosphatase activity in ATDC5 cells, ED50 ≤100 ng/ml, Bioactivity ≥1,0×10⁴ units/mg
  • Gene ID:
    O95390 / Q9Z1W4 / Q9Z217
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    BMP-11
  • Protein/peptide name:
    GDF-11
  • Purity:
    ≥95%
  • Molecular weight:
    12,5/24,9 kDa
  • Sequence:
    NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together